Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Diiodo-2-methylimidazole
4,5-Diiodo-2-methylimidazole
4,5-Diiodo-2-methylimidazole
4,5-Diiodo-2-methylimidazole
Supplier:
Apollo Scientific
Certificates
Expand image
4,5-Diiodo-2-methylimidazole
Supplier:
Apollo Scientific
About this item
CAS Number:
73746-44-8
:
Formula:
C₄H₄I₂N₂
MDL Number:
MFCD00464337
:
333.9 g/mol
Storage Temperature:
Ambient