

4,5-Dicyanoimidazole 98%
Catalog # APOSOR10446-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:118.1 g/mol
Melting Pt:168…175 °C
EINECS:214-344-6
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:1122-28-7
Formula:C₅H₂N₄
MDL Number:MFCD00005194