Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dibromo-1-methylimidazole
2,5-Dibromo-1-methylimidazole 98%
2,5-Dibromo-1-methylimidazole 98%
Irritant
2,5-Dibromo-1-methylimidazole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,5-Dibromo-1-methylimidazole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
53857-59-3
:
Formula:
C₄H₄Br₂N₂
MDL Number:
MFCD01632217
:
239.9 g/mol
Melting Pt:
68…72 °C
:
GHS07
Storage Temperature:
Refrigerator