
4-(Bromomethyl)tetrahydropyran 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:125552-89-8
:
Formula:C₆H₁₁BrO
MDL Number:MFCD03788561
:179.0549 g/mol
Boiling Pt:89…93 °C/10mmHg
Storage Temperature:Freezer
UN:3265
ADR:8,II
:GHS05