Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pyridazine-4,5-dicarboxylic acid
Pyridazine-4,5-dicarboxylic acid
Pyridazine-4,5-dicarboxylic acid
Irritant
Pyridazine-4,5-dicarboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Pyridazine-4,5-dicarboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
59648-14-5
:
Formula:
C₆H₄N₂O₄
MDL Number:
MFCD06742784
:
168.1070 g/mol
Melting Pt:
>210 °C (dec.)
:
GHS07
Storage Temperature:
Ambient