
5,6-Dihydro-4H-cyclopenta[b]thiophene-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:40133-06-0
:
Formula:C₈H₈O₂S
MDL Number:MFCD01860021
:168.22 g/mol
Melting Pt:200…202 °C
:GHS07
Storage Temperature:Ambient