Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Hydroxyindole

Irritant
Supplier:   Apollo Scientific
4-Hydroxyindole
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
Synonyms:  Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
  • CAS Number:2380-94-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00005667
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:150 °C/4mmHg
  • Melting Pt:97…99 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 49,20 €
    49,20 €
    1 * 5 g
       49,20 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 145,00 €
    145,00 €
    1 * 25 g
       145,00 €