
4-Bromothiophene-2-carbaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18791-75-8
:
Formula:C₅H₃BrOS
MDL Number:MFCD00005431
:191.05 g/mol
Boiling Pt:114…115 °C/11mmHg
Melting Pt:44…48 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator