


1-Bromo-3-chloro-5,5-dimethylhydantoin
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16079-88-2
:
Formula:C₅H₆BrClN₂O₂
MDL Number:MFCD00037350
:241.4703 g/mol
Melting Pt:160.2…161.5 °C
UN:3085
ADR:5.1+8,II
:GHS03
:GHS05
:GHS07
Storage Temperature:Ambient