
1,7-Diaminoheptane 98%
Catalog # APOSOR10104-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:130.23 g/mol
Boiling Pt:223…225 °C
Melting Pt:26…30 °C
Density:0.86 g/cm³ (25 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:211-468-2
UN:3259
ADR:8,III
:GHS05
CAS Number:646-19-5
Formula:C₇H₁₈N₂
MDL Number:MFCD00008246