Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bromocyclopentane
Bromocyclopentane
Bromocyclopentane
:
Apollo Scientific
Bromocyclopentane
:
Apollo Scientific
:
137-43-9
:
:
C₅H₉Br
:
MFCD00001359
:
149.03 g/mol
:
137…139 °C
:
1.39 g/cm³ (25 °C)
:
42 °C
:
Ambient
:
205-294-6
:
1993
:
3,III
:
GHS02