Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dibromopropane

Bottle
Catalog # APOSOR0972-50ML
Flammable
Irritant
Environment
New Product
Supplier:   Apollo Scientific
1,3-Dibromopropane
Catalog # APOSOR0972-50ML
Supplier:  Apollo Scientific
New Product
CAS Number:  109-64-8
1,3-Dibromopropane

Bottle

CAS Number:  109-64-8
Price
50,70 €
Log in for availability
1 * 50 mL
Product Details & Documents
  • CAS Number:109-64-8
  • Formula:BrH₂CCH₂CH₂Br
  • MDL Number:MFCD00000255
  • Molecular Weight:201.89 g/mol
  • Boiling Pt:167 °C
  • Melting Pt:–34 °C
  • Density:1.99 g/cm³ (25 °C)
  • Flash Pt:54 °C
  • Storage Temperature:Ambient
  • EINECS:203-690-3
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications