Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,5-dimethoxybenzaldehyde
4-Bromo-2,5-dimethoxybenzaldehyde 98%
4-Bromo-2,5-dimethoxybenzaldehyde 98%
Irritant
4-Bromo-2,5-dimethoxybenzaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Bromo-2,5-dimethoxybenzaldehyde 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
31558-41-5
:
Formula:
C₉H₉BrO₃
MDL Number:
MFCD00156964
:
245.08 g/mol
Melting Pt:
132…135 °C
:
GHS07
Storage Temperature:
Ambient