4-Hydroxy-2-methoxybenzaldehyde 99.5+%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:18278-34-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00051964
- Molecular Weight:152.15 g/mol
- Melting Pt:156…161 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:18278-34-7
- Formula:C₈H₈O₃
- MDL Number:MFCD00051964
- Molecular Weight:152.15 g/mol
- Melting Pt:156…161 °C
- Storage Temperature:Ambient



