2,3-Dimethylbenzonitrile 98%
Catalog # APOSOR0561-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:131.18 g/mol
Boiling Pt:64…66 °C/0.8mmHg
Melting Pt:23…25 °C
Storage Temperature:Ambient
CAS Number:5724-56-1
Formula:C₉H₉N
MDL Number:MFCD00019744