Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dimethyladamantane-1-carboxylic acid
3,5-Dimethyladamantane-1-carboxylic acid 95%
3,5-Dimethyladamantane-1-carboxylic acid 95%
Irritant
3,5-Dimethyladamantane-1-carboxylic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3,5-Dimethyladamantane-1-carboxylic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
14670-94-1
:
Formula:
C₁₃H₂₀O₂
MDL Number:
MFCD00188067
:
208.3 g/mol
Melting Pt:
89…90 °C
:
GHS07
Storage Temperature:
Ambient