Order Entry
Export
ContactUsLinkComponent
2,5-Dimethoxybenzenesulphonamide
Irritant
2,5-Dimethoxybenzenesulphonamide
Catalog # APOSOR0402-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,5-Dimethoxybenzenesulphonamide
Catalog # APOSOR0402-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :217.25 g/mol
Melting Pt:150…152 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:19116-90-6
Formula:C₈H₁₁NO₄S
MDL Number:MFCD00833415