Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dihydroxy-6-methylpyridine
2,4-Dihydroxy-6-methylpyridine
2,4-Dihydroxy-6-methylpyridine
Irritant
2,4-Dihydroxy-6-methylpyridine
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2,4-Dihydroxy-6-methylpyridine
Supplier:
Apollo Scientific
About this item
CAS Number:
3749-51-7
:
Formula:
C₆H₇NO₂
MDL Number:
MFCD01860849
:
125.13 g/mol
Melting Pt:
324…327 °C
:
GHS07
Storage Temperature:
Ambient