Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(3,4-Dichlorophenyl)acetohydrazide
2-(3,4-Dichlorophenyl)acetohydrazide
2-(3,4-Dichlorophenyl)acetohydrazide
Irritant
2-(3,4-Dichlorophenyl)acetohydrazide
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-(3,4-Dichlorophenyl)acetohydrazide
Supplier:
Apollo Scientific
About this item
CAS Number:
129564-33-6
:
Formula:
C₈H₈Cl₂N₂O
MDL Number:
MFCD00833407
:
219.07 g/mol
Melting Pt:
131…133 °C
:
GHS07
Storage Temperature:
Ambient