Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dichlorobenzhydrazide

3,5-Dichlorobenzhydrazide

Irritant
Supplier:   Apollo Scientific
3,5-Dichlorobenzhydrazide
Supplier:  Apollo Scientific
Certificates
3,5-Dichlorobenzhydrazide

3 Options Available Below

Product Details & Documents
Synonyms:  3,5-Dichlorobenzoic hydrazide
  • CAS Number:62899-78-9
  • Formula:C₇H₆Cl₂N₂O
  • MDL Number:MFCD00833411
  • Molecular Weight:205.04 g/mol
  • Melting Pt:205…207 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 43,50 €
    43,50 €
    1 * 1 g
       43,50 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 145,00 €
    145,00 €
    1 * 5 g
       145,00 €