Order Entry
Export
ContactUsLinkComponent
3,5-Dichlorobenzenesulphonamide 95+%
Irritant
3,5-Dichlorobenzenesulphonamide 95+%
Catalog # APOSOR0340-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,5-Dichlorobenzenesulphonamide 95+%
Catalog # APOSOR0340-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.08 g/mol
Melting Pt:161…165 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:19797-32-1
Formula:C₆H₅Cl₂NO₂S
MDL Number:MFCD00117161