Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Bis(acetamido)benzoic acid
3,4-Bis(acetamido)benzoic acid
3,4-Bis(acetamido)benzoic acid
APOSOR0327-1G
:
Apollo Scientific
:
3,4-Bis(acetamido)benzoic acid
APOSOR0327-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
236.23 g/mol
:
226…228 °C
:
GHS07
:
Ambient
:
205873-56-9
:
C₁₁H₁₂N₂O₄
:
MFCD00238577