Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Cyclohexanedione
1,2-Cyclohexanedione 95%
1,2-Cyclohexanedione 95%
:
Apollo Scientific
1,2-Cyclohexanedione 95%
:
Apollo Scientific
:
765-87-7
:
:
193…195 °C
:
34…38 °C
:
1.12 g/cm³ (25 °C)
:
212-155-3