
3-Chloro-2,5-dihydrothiophene-1,1-dioxide
Catalog # APOSOR0299-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:152.6 g/mol
Melting Pt:98…102 °C
:GHS07
Storage Temperature:Ambient
CAS Number:7311-87-7
Formula:C₄H₅ClO₂S
MDL Number:MFCD00154884