
4-(Chloromethyl)-3,5-dimethylisoxazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:19788-37-5
:
Formula:C₆H₈ClNO
MDL Number:MFCD00003154
Boiling Pt:88…90 °C/7mmHg
Density:1.167 g/cm³ (25 °C)
Flash Pt:95 °C
:145.59 g/mol
EINECS:243-312-4
:GHS07
Storage Temperature:Refrigerator