Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(5-Chloropyridin-2-yl)-1,4-diazepane
1-(5-Chloropyridin-2-yl)-1,4-diazepane
1-(5-Chloropyridin-2-yl)-1,4-diazepane
:
Apollo Scientific
1-(5-Chloropyridin-2-yl)-1,4-diazepane
:
Apollo Scientific
:
502133-62-2
:
:
C₁₀H₁₄ClN₃
:
MFCD05182308
:
211.7 g/mol
:
GHS07
:
Ambient