Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 2,6-dichlorobenzoate
Ethyl-2,6-dichlorobenzoate
Ethyl-2,6-dichlorobenzoate
Irritant
Ethyl-2,6-dichlorobenzoate
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Ethyl-2,6-dichlorobenzoate
Supplier:
Apollo Scientific
About this item
CAS Number:
81055-73-4
:
Formula:
C₉H₈Cl₂O₂
MDL Number:
MFCD01313702
:
219.07 g/mol
Boiling Pt:
264…265 °C
:
GHS07
Storage Temperature:
Ambient