Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,6-Naphthyridine-3-carboxylic acid
1,6-Naphthyridine-3-carboxylic acid 95+%
1,6-Naphthyridine-3-carboxylic acid 95+%
Irritant
1,6-Naphthyridine-3-carboxylic acid 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,6-Naphthyridine-3-carboxylic acid 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
1017793-59-7
:
Formula:
C₉H₆N₂O₂
MDL Number:
MFCD07366468
:
174.16 g/mol
Melting Pt:
193 °C
:
GHS07
Storage Temperature:
Ambient