Order Entry
Export
Orders LinkContactUsLinkComponent
 

2'-Hydroxy-4'-methoxyacetophenone

Irritant
Supplier:   Apollo Scientific
2'-Hydroxy-4'-methoxyacetophenone
Supplier:  Apollo Scientific
Certificates
2'-Hydroxy-4'-methoxyacetophenone

6 Options Available Below

Product Details & Documents
Synonyms:  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  • CAS Number:552-41-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00008730
  • Molecular Weight:166.18 g/mol
  • Melting Pt:48…50 °C
  • Flash Pt:113 °C
  • Storage Temperature:Refrigerator
  • EINECS:209-012-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 23,20 €
    23,20 €
    1 * 25 g
       23,20 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 56,50 €
    56,50 €
    1 * 100 g
       56,50 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 92,70 €
    92,70 €
    1 * 500 g
       92,70 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 452,00 €
    452,00 €
    1 * 2,5 kg
       452,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 10 kg
    1 * 10 kg - 1 790,00 €
    1 790,00 €
    1 * 10 kg
       1 790,00 €

    Specifications:
    New Product
    New Product