Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ω-Bromo-2,4-dimethylacetophenone
ω-Bromo-2,4-dimethylacetophenone 97%
ω-Bromo-2,4-dimethylacetophenone 97%
Corrosive
ω-Bromo-2,4-dimethylacetophenone 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
ω-Bromo-2,4-dimethylacetophenone 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
26346-85-0
:
Formula:
C₁₀H₁₁BrO
MDL Number:
MFCD00017872
:
227.1
Melting Pt:
43…45 °C
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III
:
GHS05