
2,4'-Bipyridyl 97%
Catalog # APOSOR015006-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:156.19 g/mol
Boiling Pt:280…282 °C/760mmHg
Melting Pt:58…62 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:581-47-5
Formula:C₁₀H₈N₂
MDL Number:MFCD00006217