Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-4,5-dimethylthiazole hydrochloride
2-Amino-4,5-dimethylthiazole hydrochloride 98%
2-Amino-4,5-dimethylthiazole hydrochloride 98%
:
Apollo Scientific
2-Amino-4,5-dimethylthiazole hydrochloride 98%
:
Apollo Scientific
:
71574-33-9
:
:
C₅H₉ClN₂S
:
MFCD00012711
:
164.66 g/mol
:
273…274 °C (dec.)
:
GHS07
:
Ambient