Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichlorosalicylaldehyde
4,6-Dichlorosalicylaldehyde 98%
4,6-Dichlorosalicylaldehyde 98%
Irritant
4,6-Dichlorosalicylaldehyde 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4,6-Dichlorosalicylaldehyde 98%
Supplier:
Apollo Scientific
Certificates
About this item
CAS Number:
78443-72-8
:
Formula:
C₇H₄Cl₂O₂
MDL Number:
MFCD00016590
:
191.01 g/mol
Melting Pt:
50…53 °C
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
Documentation