Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate 95%
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate 95%
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate 95%
APOSOR01483-500MG
:
Apollo Scientific
:
Methyl-2,4-dioxo-4-pyridin-3-ylbutanoate 95%
APOSOR01483-500MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 mg
Seq:
2725
:
:
207.19 g/mol
:
119 °C
:
GHS07
:
Ambient
:
23424-36-4
:
C₁₀H₉NO₄
:
MFCD06245328