Order Entry
Export
Orders LinkContactUsLinkComponent
3,4-Dichlorophenol 98%

3,4-Dichlorophenol 98%

Irritant
Corrosive
Environment
Supplier:   Apollo Scientific
3,4-Dichlorophenol 98%
Supplier:  Apollo Scientific
Certificates
3,4-Dichlorophenol 98%

2 Options Available Below

Product Details & Documents
  • CAS Number:95-77-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002258
  • Molecular Weight:163 g/mol
  • Boiling Pt:145…146 °C
  • Melting Pt:64…68 °C
  • Storage Temperature:Ambient
  • EINECS:202-450-5
  • UN:2020
  • ADR:6.1,III
  •  :Density: 1.6 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 142,00 €
    142,00 €
    1 * 100 g
       142,00 €

  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 225,00 €
    225,00 €
    1 * 250 g
       225,00 €