Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Diaminopyridine

2,3-Diaminopyridine

Toxic
New Product
Supplier:   Apollo Scientific
2,3-Diaminopyridine
Supplier:  Apollo Scientific
New Product
Certificates
2,3-Diaminopyridine

2 Options Available Below

Product Details & Documents
Synonyms:  Pyridine-2,3-diyldiamine
  • CAS Number:452-58-4
  • Formula:C₅H₇N₃
  • MDL Number:MFCD00006319
  • Molecular Weight:109.13 g/mol
  • Boiling Pt:195 °C/55mmHg
  • Melting Pt:110…115 °C
  • Storage Temperature:Ambient
  • EINECS:207-200-9
  • UN:2811
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 228,00 €
    228,00 €
    1 * 100 g
       228,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 030,00 €
    1 030,00 €
    1 * 500 g
       1 030,00 €

    Specifications:
    New Product
    New Product