Order Entry
Export
Orders LinkContactUsLinkComponent
Zinc molybdate

Zinc molybdate

Irritant
Supplier:   Apollo Scientific
Zinc molybdate
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Zinc molybdenum oxide
  • CAS Number:13767-32-3
  • Formula:MoO₄Zn
  • MDL Number:MFCD00049622
  • Molecular Weight:225.326 g/mol
  • Melting Pt:>700 °C
  • Melting Pt:>700 °C
  • Density:4.3 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:237-377-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 108,00 €
    108,00 €
    1 * 5 g
       108,00 €

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 387,00 €
    387,00 €
    1 * 25 g
       387,00 €