Tungsten trioxide 99+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1314-35-8
:
Formula:O₃W
MDL Number:MFCD00011466
:231.84 g/mol
Melting Pt:1473 °C
Density:7.16 g/cm³ (25 °C)
EINECS:215-231-4
:GHS07
Storage Temperature:Ambient