

Terbium 99,9%, powder –40 mesh
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7440-27-9
:
Formula:Tb
MDL Number:MFCD00011256
:158.93 g/mol
Boiling Pt:2800 °C
Melting Pt:1356 °C
Density:8.27 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3178
ADR:4.1,II
:GHS02
:GHS07