Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silver(I) oxide
Silver(I) oxide 99,5%
Silver(I) oxide 99,5%
Silver(I) oxide 99,5%
APOSIN3238-1G
:
Apollo Scientific
:
Silver(I) oxide 99,5%
APOSIN3238-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
231.74 g/mol
:
230 °C (dec.)
:
7.2 g/cm³ (25 °C)
:
Ambient
:
243-957-1
:
1479
:
5.1,II
:
GHS03
:
GHS05
:
GHS09
:
20667-12-3
:
Ag₂O
:
MFCD00003404