Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) bromide 99.999%

Lead(II) bromide 99.999%

Bottle
Catalog # APOSIN2209-25G
Irritant
Health
Environment
New Product
Supplier:   Apollo Scientific
Lead(II) bromide 99.999%
Catalog # APOSIN2209-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  10031-22-8
Lead(II) bromide 99.999%

Bottle

CAS Number:  10031-22-8
Price
427,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Lead dibromide, Lead(II) bromide
  • CAS Number:10031-22-8
  • Formula:Br₂Pb
  • MDL Number:MFCD00011156
  • Molecular Weight:367.01 g/mol
  • Boiling Pt:916 °C
  • Melting Pt:373 °C
  • Density:6.66 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:233-084-4
  • UN:2291
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications