
Iron(II) acetate, anhydrous
Catalog # APOSIN2041-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:173.94 g/mol
Melting Pt:190…200 °C (dec.)
EINECS:221-441-7
:GHS07
Storage Temperature:Ambient
CAS Number:3094-87-9
Formula:C₄H₆FeO₄
MDL Number:MFCD00058909