Order Entry
Export
ContactUsLinkComponent
Copper(II) acetate monohydrate 98%
Irritant
Environment
Copper(II) acetate monohydrate 98%
Catalog # APOSIN1533-1KG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
Copper(II) acetate monohydrate 98%
Catalog # APOSIN1533-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Melting Pt:115 °C
Density:1.882 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
 :199.65 g/mol
EINECS:205-553-3
 :GHS07
 :GHS09
CAS Number:6046-93-1
Formula:(H₃CCOO)₂Cu·H₂O
MDL Number:MFCD00149570