Order Entry
Export
ContactUsLinkComponent
Copper 99.999%, shot 4 - 6 mm
Irritant
Environment
Copper 99.999%, shot 4 - 6 mm
Catalog # APOSIN1531-250G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
Copper 99.999%, shot 4 - 6 mm
Catalog # APOSIN1531-250G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :63.55 g/mol
Boiling Pt:2595 °C
Melting Pt:1083 °C
Density:8.94 g/cm³ (25 °C)
EINECS:231-159-6
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:7440-50-8
Formula:Cu
MDL Number:MFCD00010965