
Aluminium chloride, anhydrous 99.99+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7446-70-0
:
Formula:AlCl₃
MDL Number:MFCD00003422
:133.34 g/mol
Melting Pt:190 °C
Density:2.44 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-208-1
UN:1726
ADR:8,II
:GHS05