


Potassium tetrachloroplatinate(II) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10025-99-7
:
Formula:Cl₄K₂Pt
MDL Number:MFCD00011378
:415.09 g/mol
Density:3.38 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-050-9
UN:3288
ADR:6.1,III
:GHS05
:GHS06
:GHS08