Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PX 12
PX 12
PX 12
PX 12
Supplier:
Apollo Scientific
Certificates
Expand image
PX 12
Supplier:
Apollo Scientific
About this item
CAS Number:
141400-58-0
:
Formula:
C₇H₁₂N₂S₂
MDL Number:
MFCD18086851
:
188.3136 g/mol
Storage Temperature:
Ambient