AGI-6780
New Product
Supplier: Apollo Scientific
Jump to:
About this item
AGI-6780 is a selective inhibitor of IDH2R140Q mutant with IC50 value of 23 nM.
Synonyms: N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide
- CAS Number:1432660-47-3
- Formula:C₂₁H₁₈F₃N₃O₃S₂
- MDL Number:MFCD26097285
- Molecular Weight:481.51 g/mol
- Storage Temperature:Ambient
4 Options Available Below
Product Details & Documents
AGI-6780 is a selective inhibitor of IDH2R140Q mutant with IC50 value of 23 nM.
Synonyms: N-Cyclopropyl-4-(thiophen-3-yl)-3-(3-(3-(trifluoromethyl)phenyl)ureido)benzenesulphonamide
- CAS Number:1432660-47-3
- Formula:C₂₁H₁₈F₃N₃O₃S₂
- MDL Number:MFCD26097285
- Molecular Weight:481.51 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



