Order Entry
Export
ContactUsLinkComponent
JC-1 iodide salt 99%
JC-1 iodide salt 99%
Catalog # APOSBISN0055-5MG
Supplier:  Apollo Scientific
CAS Number:  
JC-1 iodide salt 99%
Catalog # APOSBISN0055-5MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :652.23 g/mol
Storage Temperature:Ambient
CAS Number:47729-63-5
 :3520-43-2
Formula:C₂₅H₂₇Cl₄IN₄
MDL Number:MFCD00188475