
21-[(tert-Butoxycarbonyl)amino]-4,7,10,13,16,19-hexaoxaheneicosanoic acid
Catalog # APOSBIPG1804-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:453.53 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:882847-13-4
Formula:C₂₀H₃₉NO₁₀
MDL Number:MFCD07781255